Assay Details
| Gene Symbol: cdh-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066286.6 | NP_498687.2 | 9 - 10 | 2136 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdh-3| Gene Symbol: cdh-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066286.6 | NP_498687.2 | 9 - 10 | 2136 | 110 | |
Set Membership: |
|