Assay Details
Gene Symbol: ZC262.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306707.1 | NP_001293636.1 | 1 - 2 | 135 | 59 |
Gene Symbol: R05D3.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382936.1 | 6 - 7 | 2231 | 59 |
Gene Symbol: unc-116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382938.1 | 7 - 8 | 2362 | 59 | ||
GenBank mRNA | AB017163.1 | - | 6 - 7 | 2220 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382936.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
