Assay Details
| Gene Symbol: polk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129220.3 | NP_001122692.1 | 4 - 5 | 1038 | 71 | |
| NM_001382942.1 | 4 - 5 | 988 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

polk-1| Gene Symbol: polk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129220.3 | NP_001122692.1 | 4 - 5 | 1038 | 71 | |
| NM_001382942.1 | 4 - 5 | 988 | 71 | |||
Set Membership: |
|