Assay Details
| Gene Symbol: fli-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066512.6 | NP_498913.2 | 10 - 11 | 3213 | 93 | |
| GenBank mRNA | U01183.1 | - | 11 - 12 | 3246 | 93 | |
More Information
Set Membership: |
|

fli-1
Genomic Map