Assay Details
Gene Symbol: fli-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066512.6 | NP_498913.2 | 11 - 12 | 3565 | 92 | |
GenBank mRNA | U01183.1 | - | 12 - 13 | 3598 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
