Assay Details
Gene Symbol: fli-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066512.6 | NP_498913.2 | 6 - 7 | 1243 | 104 | |
GenBank mRNA | U01183.1 | - | 7 - 8 | 1276 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
