Assay Details
| Gene Symbol: rpn-6.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268043.2 | NP_001254972.1 | 2 - 3 | 552 | 70 | |
| Gene Symbol: F59B2.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379854.1 | NP_001366671.1 | 2 - 3 | 564 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379854.1 transcripts.
Set Membership: |
|

rpn-6.2
Genomic Map