Assay Details
Gene Symbol: nac-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268047.2 | NP_001254976.1 | 12 - 13 | 1762 | 99 | |
NM_001268048.2 | NP_001254977.1 | 11 - 12 | 1509 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
