Assay Details
Gene Symbol: nac-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268047.2 | NP_001254976.1 | 4 - 5 | 602 | 88 | |
NM_001268048.2 | NP_001254977.1 | 3 - 4 | 349 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
