Assay Details
Gene Symbol: nac-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268047.2 | NP_001254976.1 | 5 - 6 | 641 | 64 | |
NM_001268048.2 | NP_001254977.1 | 4 - 5 | 388 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
