Assay Details
| Gene Symbol: sec-16A.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306722.3 | NP_001293651.1 | 5 - 6 | 1012 | 59 | |
| NM_066621.5 | NP_499022.1 | 5 - 6 | 1036 | 59 | ||
More Information
Set Membership: |
|

sec-16A.1
Genomic Map