Assay Details
Gene Symbol: clip-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027511.6 | NP_001022682.1 | 14 - 15 | 1867 | 76 | |
NM_001027512.7 | NP_001022683.1 | 13 - 14 | 1663 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
