Assay Details
| Gene Symbol: emb-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027491.7 | NP_001022662.2 | 7 - 8 | 2075 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

emb-9| Gene Symbol: emb-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027491.7 | NP_001022662.2 | 7 - 8 | 2075 | 73 | |
Set Membership: |
|