Assay Details
| Gene Symbol: emb-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066673.7 | NP_499074.2 | 3 - 4 | 509 | 106 | |
| GenBank mRNA | AF192400.1 | - | 3 - 4 | 428 | 106 | |
More Information
Set Membership: |
|

emb-30
Genomic Map