Assay Details
| Gene Symbol: bre-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066686.7 | NP_499087.2 | 2 - 3 | 246 | 82 | |
| NR_132454.1 | 2 - 3 | 224 | 82 | |||
| GenBank mRNA | AY533305.1 | - | 1 - 2 | 131 | 82 | |
More Information
Set Membership: |
|

bre-3
Genomic Map