Assay Details
Gene Symbol: tlk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382967.1 | 7 - 8 | 2915 | 85 | ||
NM_066744.5 | NP_499145.2 | 8 - 9 | 2985 | 85 | ||
GenBank mRNA | AY450852.1 | - | 7 - 8 | 2797 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
