Assay Details
| Gene Symbol: cbp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066759.5 | NP_499160.2 | 10 - 11 | 5861 | 72 | |
| NM_066760.7 | NP_499161.2 | 10 - 11 | 5822 | 72 | ||
More Information
Set Membership: |
|

cbp-1
Genomic Map