Assay Details
| Gene Symbol: cyld-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268116.2 | NP_001255045.1 | 10 - 11 | 1205 | 159 | |
| NM_001268117.3 | NP_001255046.1 | 4 - 5 | 457 | 159 | ||
| NM_001268118.3 | NP_001255047.1 | 2 - 3 | 274 | 159 | ||
More Information
Set Membership: |
|

cyld-1
Genomic Map