Assay Details
| Gene Symbol: cyld-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268116.2 | NP_001255045.1 | 11 - 12 | 1350 | 147 | |
| NM_001268117.3 | NP_001255046.1 | 5 - 6 | 602 | 147 | ||
| NM_001268118.3 | NP_001255047.1 | 3 - 4 | 419 | 147 | ||
More Information
Set Membership: |
|

cyld-1
Genomic Map