Assay Details
| Gene Symbol: cyld-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268116.2 | NP_001255045.1 | 5 - 6 | 643 | 155 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cyld-1| Gene Symbol: cyld-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268116.2 | NP_001255045.1 | 5 - 6 | 643 | 155 | |
Set Membership: |
|