Assay Details
| Gene Symbol: mog-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066811.9 | NP_499212.1 | 6 - 7 | 3078 | 113 | |
| GenBank mRNA | AF120269.1 | - | 6 - 7 | 3093 | 113 | |
More Information
Set Membership: |
|

mog-1
Genomic Map