Assay Details
| Gene Symbol: rgs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268127.3 | NP_001255056.1 | 1 - 2 | 503 | 77 | |
| NM_001382973.1 | 1 - 2 | 527 | 77 | |||
| GenBank mRNA | AF220159.1 | - | 1 - 2 | 548 | 77 | |
More Information
Set Membership: |
|

rgs-1
Genomic Map