Assay Details
Gene Symbol: rgs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382973.1 | 3 - 4 | 675 | 158 | ||
GenBank mRNA | AF220159.1 | - | 3 - 4 | 696 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
