Assay Details
| Gene Symbol: rgs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382973.1 | 4 - 5 | 731 | 109 | ||
| GenBank mRNA | AF220159.1 | - | 4 - 5 | 752 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rgs-1| Gene Symbol: rgs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382973.1 | 4 - 5 | 731 | 109 | ||
| GenBank mRNA | AF220159.1 | - | 4 - 5 | 752 | 109 | |
Set Membership: |
|