Assay Details
| Gene Symbol: T16G12.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379878.2 | NP_001366683.1 | 7 - 8 | 1356 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

T16G12.6| Gene Symbol: T16G12.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379878.2 | NP_001366683.1 | 7 - 8 | 1356 | 85 | |
Set Membership: |
|