Assay Details
Gene Symbol: mua-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027503.4 | NP_001022674.1 | 14 - 15 | 2081 | 85 | |
NM_001027504.3 | NP_001022675.1 | 4 - 5 | 721 | 85 | ||
GenBank mRNA | AF139060.1 | - | 14 - 15 | 2081 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
