Assay Details
| Gene Symbol: mua-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027503.4 | NP_001022674.1 | 4 - 5 | 731 | 60 | |
| GenBank mRNA | AF139060.1 | - | 4 - 5 | 731 | 60 | |
More Information
Set Membership: |
|

mua-3
Genomic Map