Assay Details
Gene Symbol: chc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066859.5 | NP_499260.1 | 1 - 2 | 171 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
