Assay Details
| Gene Symbol: T20G5.9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066860.3 | NP_499261.2 | 3 - 4 | 293 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

T20G5.9| Gene Symbol: T20G5.9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066860.3 | NP_499261.2 | 3 - 4 | 293 | 65 | |
Set Membership: |
|