Assay Details
| Gene Symbol: cnk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066873.6 | NP_499274.3 | 4 - 5 | 747 | 81 | |
| GenBank mRNA | DQ104391.1 | - | 4 - 5 | 747 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cnk-1| Gene Symbol: cnk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066873.6 | NP_499274.3 | 4 - 5 | 747 | 81 | |
| GenBank mRNA | DQ104391.1 | - | 4 - 5 | 747 | 81 | |
Set Membership: |
|