Assay Details
| Gene Symbol: cnk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066873.6 | NP_499274.3 | 8 - 9 | 1749 | 65 | |
| GenBank mRNA | DQ104391.1 | - | 8 - 9 | 1749 | 65 | |
More Information
Set Membership: |
|

cnk-1
Genomic Map