Assay Details
Gene Symbol: prp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066899.8 | NP_499300.1 | 3 - 4 | 1443 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
