Assay Details
Gene Symbol: atx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379897.1 | NP_001367241.1 | 1 - 2 | 340 | 61 | |
NM_001379898.1 | NP_001366696.1 | 1 - 2 | 101 | 61 | ||
NM_001379900.2 | NP_001367113.1 | 1 - 2 | 83 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379898.1, NM_001379900.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
