Assay Details
Gene Symbol: atx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379897.1 | NP_001367241.1 | 4 - 5 | 1065 | 65 | |
NM_001379898.1 | NP_001366696.1 | 4 - 5 | 826 | 65 | ||
NM_001379899.1 | NP_001367186.1 | 3 - 4 | 741 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
