Assay Details
| Gene Symbol: unc-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027611.3 | NP_001022782.1 | 11 - 12 | 1411 | 69 | |
| NM_001306733.3 | NP_001293662.1 | 1 - 2 | 52 | 69 | ||
| NM_001379903.1 | NP_001368253.1 | 7 - 8 | 784 | 69 | ||
| GenBank mRNA | AF151640.1 | - | 11 - 12 | 1580 | 69 | |
More Information
Set Membership: |
|

unc-49
Genomic Map