Assay Details
| Gene Symbol: unc-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027611.3 | NP_001022782.1 | 12 - 13 | 1491 | 75 | |
| NM_001306733.3 | NP_001293662.1 | 2 - 3 | 132 | 75 | ||
| NM_001379903.1 | NP_001368253.1 | 8 - 9 | 864 | 75 | ||
| NM_001382981.1 | 9 - 10 | 1318 | 75 | |||
| GenBank mRNA | AF151640.1 | - | 12 - 13 | 1660 | 75 | |
More Information
Set Membership: |
|

unc-49
Genomic Map