Assay Details
| Gene Symbol: unc-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027611.3 | NP_001022782.1 | 1 - 2 | 195 | 70 | |
| NM_001027612.4 | NP_001022783.1 | 1 - 2 | 278 | 70 | ||
| NM_001027613.3 | NP_001022784.1 | 1 - 2 | 278 | 70 | ||
| NM_001027614.4 | NP_001022785.1 | 1 - 2 | 197 | 70 | ||
| NM_001027615.4 | NP_001022786.1 | 1 - 2 | 278 | 70 | ||
| NM_001379901.2 | NP_001368252.1 | 1 - 2 | 197 | 70 | ||
| NM_001379904.1 | NP_001367142.1 | 1 - 2 | 278 | 70 | ||
| NM_001382981.1 | 1 - 2 | 244 | 70 | |||
| GenBank mRNA | AF151640.1 | - | 1 - 2 | 364 | 70 | |
| AF151641.1 | - | 1 - 2 | 204 | 70 | ||
| AF151642.1 | - | 1 - 2 | 204 | 70 | ||
| AF151643.1 | - | 1 - 2 | 204 | 70 | ||
| AF151644.1 | - | 1 - 2 | 185 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379904.1 transcripts.
Set Membership: |
|

unc-49
Genomic Map