Assay Details
| Gene Symbol: unc-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027611.3 | NP_001022782.1 | 17 - 18 | 2312 | 58 | |
| NM_001027612.4 | NP_001022783.1 | 10 - 11 | 1495 | 58 | ||
| NM_001027613.3 | NP_001022784.1 | 9 - 10 | 1459 | 58 | ||
| NM_001027614.4 | NP_001022785.1 | 11 - 12 | 1471 | 58 | ||
| NM_001379901.2 | NP_001368252.1 | 10 - 11 | 1414 | 58 | ||
| NM_001379902.1 | NP_001368254.1 | 6 - 7 | 785 | 58 | ||
| NM_001379904.1 | NP_001367142.1 | 10 - 11 | 1495 | 58 | ||
| NM_001382981.1 | 14 - 15 | 2310 | 58 | |||
| GenBank mRNA | AF151641.1 | - | 10 - 11 | 1421 | 58 | |
| AF151642.1 | - | 9 - 10 | 1385 | 58 | ||
| AF151643.1 | - | 11 - 12 | 1478 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379904.1 transcripts.
Set Membership: |
|

unc-49
Genomic Map