Assay Details
| Gene Symbol: unc-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027611.3 | NP_001022782.1 | 18 - 19 | 2401 | 87 | |
| NM_001027612.4 | NP_001022783.1 | 11 - 12 | 1584 | 87 | ||
| NM_001027613.3 | NP_001022784.1 | 10 - 11 | 1548 | 87 | ||
| NM_001027614.4 | NP_001022785.1 | 12 - 13 | 1560 | 87 | ||
| NM_001379901.2 | NP_001368252.1 | 11 - 12 | 1503 | 87 | ||
| NM_001379902.1 | NP_001368254.1 | 7 - 8 | 874 | 87 | ||
| NM_001379904.1 | NP_001367142.1 | 11 - 12 | 1584 | 87 | ||
| NM_001382981.1 | 15 - 16 | 2399 | 87 | |||
| GenBank mRNA | AF151641.1 | - | 11 - 12 | 1510 | 87 | |
| AF151642.1 | - | 10 - 11 | 1474 | 87 | ||
| AF151643.1 | - | 12 - 13 | 1567 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379904.1 transcripts.
Set Membership: |
|

unc-49
Genomic Map