Assay Details
| Gene Symbol: unc-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027611.3 | NP_001022782.1 | 16 - 17 | 2184 | 88 | |
| NM_001027613.3 | NP_001022784.1 | 8 - 9 | 1331 | 88 | ||
| GenBank mRNA | AF151642.1 | - | 8 - 9 | 1257 | 88 | |
More Information
Set Membership: |
|

unc-49
Genomic Map