Assay Details
Gene Symbol: tat-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066962.5 | NP_499363.3 | 5 - 6 | 1787 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
