Assay Details
Gene Symbol: sca-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066984.8 | NP_499385.3 | 3 - 4 | 153 | 79 | |
NM_066985.6 | NP_499386.1 | 3 - 4 | 199 | 79 | ||
GenBank mRNA | AJ012296.1 | - | 2 - 3 | 173 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
