Assay Details
Gene Symbol: sca-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129236.4 | NP_001122708.1 | 1 - 2 | 90 | 85 | |
NM_066984.8 | NP_499385.3 | 5 - 6 | 507 | 85 | ||
NM_066985.6 | NP_499386.1 | 5 - 6 | 553 | 85 | ||
GenBank mRNA | AJ012296.1 | - | 4 - 5 | 527 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
