Assay Details
Gene Symbol: sca-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129236.4 | NP_001122708.1 | 2 - 3 | 2152 | 86 | |
NM_066984.8 | NP_499385.3 | 6 - 7 | 2569 | 86 | ||
NM_066985.6 | NP_499386.1 | 6 - 7 | 2615 | 86 | ||
GenBank mRNA | AJ012296.1 | - | 5 - 6 | 2589 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
