Assay Details
| Gene Symbol: unc-119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268159.3 | NP_001255088.1 | 2 - 3 | 145 | 66 | |
| NM_001268160.4 | NP_001255089.1 | 1 - 2 | 77 | 66 | ||
More Information
Set Membership: |
|

unc-119
Genomic Map