Assay Details
| Gene Symbol: Y47D3A.14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067044.5 | NP_499445.2 | 2 - 3 | 171 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Y47D3A.14| Gene Symbol: Y47D3A.14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067044.5 | NP_499445.2 | 2 - 3 | 171 | 86 | |
Set Membership: |
|