Assay Details
| Gene Symbol: smc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268189.3 | NP_001255118.1 | 15 - 16 | 3084 | 65 | |
| NM_001268190.3 | NP_001255119.1 | 2 - 3 | 425 | 65 | ||
More Information
Set Membership: |
|

smc-3
Genomic Map