Assay Details
| Gene Symbol: smc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268189.3 | NP_001255118.1 | 18 - 19 | 3469 | 69 | |
| NM_001268190.3 | NP_001255119.1 | 5 - 6 | 810 | 69 | ||
More Information
Set Membership: |
|

smc-3
Genomic Map