Assay Details
| Gene Symbol: subs-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067064.4 | NP_499465.3 | 10 - 11 | 1553 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

subs-4| Gene Symbol: subs-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067064.4 | NP_499465.3 | 10 - 11 | 1553 | 100 | |
Set Membership: |
|