Assay Details
Gene Symbol: spe-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067081.4 | NP_499482.1 | 3 - 4 | 350 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
